Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
ENST00000579506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
ENST00000579506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
ENST00000581198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
ENST00000581198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
ENST00000581198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
ENST00000581198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
ENST00000581198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
ENST00000581198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
NR_110798.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
NR_110798.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
NR_110798.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
NR_110798.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
NR_110798.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
NR_110798.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007345.hg.1 |
NR_110798.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150323.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150323.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150323.hg.1 |
NR_110798.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150323.hg.1 |
TC1800007345.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800150323.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150323.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150323.hg.1 |
NR_110798.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
ENST00000578610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
ENST00000579506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
ENST00000579506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
ENST00000581198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
ENST00000581198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
ENST00000581198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
ENST00000581198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
ENST00000581198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
ENST00000581198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
NR_110798.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
NR_110798.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
NR_110798.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
NR_110798.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
NR_110798.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
NR_110798.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007345.hg.1 |
NR_110798.1 |
Refseq |