Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008160.hg.1 |
ENST00000579972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008160.hg.1 |
ENST00000579972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008160.hg.1 |
ENST00000579972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008160.hg.1 |
NR_110804.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008160.hg.1 |
NR_110804.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008160.hg.1 |
NR_110804.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008160.hg.1 |
NR_110804.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008160.hg.1 |
NR_110804.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008160.hg.1 |
NR_110804.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008160.hg.1 |
NR_110804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160261.hg.1 |
NR_110804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160261.hg.1 |
NR_110804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160261.hg.1 |
TC1700008160.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700008160.hg.1 |
ENST00000579972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008160.hg.1 |
ENST00000579972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008160.hg.1 |
ENST00000579972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008160.hg.1 |
NR_110804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008160.hg.1 |
NR_110804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008160.hg.1 |
NR_110804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008160.hg.1 |
NR_110804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008160.hg.1 |
NR_110804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008160.hg.1 |
NR_110804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008160.hg.1 |
NR_110804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160265.hg.1 |
ENST00000579972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160265.hg.1 |
NR_110804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160265.hg.1 |
ENST00000579972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160265.hg.1 |
NR_110804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160265.hg.1 |
TC1700008160.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700160266.hg.1 |
ENST00000579972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160266.hg.1 |
NR_110804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160266.hg.1 |
ENST00000579972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160266.hg.1 |
NR_110804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160266.hg.1 |
TC1700008160.hg.1 |
Clariom D TC |