Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011222.hg.1 |
ENST00000577189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011222.hg.1 |
ENST00000577189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011222.hg.1 |
ENST00000577189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011222.hg.1 |
NR_110808.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011222.hg.1 |
NR_110808.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011222.hg.1 |
NR_110808.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011222.hg.1 |
ENST00000577189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011222.hg.1 |
ENST00000577189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011222.hg.1 |
ENST00000577189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011222.hg.1 |
NR_110808.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011222.hg.1 |
NR_110808.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011222.hg.1 |
NR_110808.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190144.hg.1 |
ENST00000577189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190144.hg.1 |
NR_110808.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190144.hg.1 |
ENST00000577189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190144.hg.1 |
NR_110808.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190144.hg.1 |
TC1700011222.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700190143.hg.1 |
ENST00000577189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190143.hg.1 |
NR_110808.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190143.hg.1 |
ENST00000577189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190143.hg.1 |
NR_110808.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190143.hg.1 |
TC1700011222.hg.1 |
Clariom D TC |