Assay Details
| Gene Symbol: LOC101928700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110829.1 | 2 - 3 | 312 | 129 | ||
| GenBank mRNA | BC026117.1 | - | 2 - 3 | 317 | 129 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | thiamin pyrophosphokinase 1 | TPK1 |
HTPK1, PP20, THMD5 |
-2.2mb | Sense | Chr. 7 - 144451941 - 144836424 on NCBI Build GRCh38 |
| Downstream | microRNA 548f-4 | MIR548F4 |
MIR548F-4, MIRN548F4, hsa-mir-548f-4 |
292.2kb | Sense | Chr. 7 - 147378017 - 147378121 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101928700
Genomic Map