| Array Name | 
									Probe IDs | 
									Targets | 
									Target Type | 
								
	
								
									
										
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1700009048.hg.1 | 
												ENST00000587575.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1700009048.hg.1 | 
												ENST00000587575.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1700009048.hg.1 | 
												ENST00000587575.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1700009048.hg.1 | 
												ENST00000592569.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1700009048.hg.1 | 
												ENST00000592569.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1700009048.hg.1 | 
												ENST00000592569.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1700009048.hg.1 | 
												ENST00000592569.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1700009048.hg.1 | 
												ENST00000592569.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1700009048.hg.1 | 
												NR_110845.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1700009048.hg.1 | 
												NR_110845.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1700009048.hg.1 | 
												NR_110845.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1700009048.hg.1 | 
												NR_110845.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1700009048.hg.1 | 
												NR_110845.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1700009048.hg.1 | 
												NR_110845.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1700009048.hg.1 | 
												NR_110846.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1700009048.hg.1 | 
												NR_110846.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1700009048.hg.1 | 
												NR_110846.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1700009048.hg.1 | 
												NR_110846.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1700009048.hg.1 | 
												NR_110846.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1700009048.hg.1 | 
												NR_110847.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1700009048.hg.1 | 
												NR_110847.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1700009048.hg.1 | 
												NR_110847.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1700009048.hg.1 | 
												NR_110847.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1700009048.hg.1 | 
												NR_110847.1 | 
												Refseq | 
												
											
										
										
									
								
									
										
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1700168377.hg.1 | 
												ENST00000592569.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1700168377.hg.1 | 
												NR_110845.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1700168377.hg.1 | 
												NR_110847.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1700168377.hg.1 | 
												ENST00000592569.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1700168377.hg.1 | 
												NR_110845.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1700168377.hg.1 | 
												NR_110847.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1700168377.hg.1 | 
												TC1700009048.hg.1 | 
												Clariom D TC | 
												
											
										
										
									
								
									
										
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1700009048.hg.1 | 
												ENST00000587575.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1700009048.hg.1 | 
												ENST00000587575.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1700009048.hg.1 | 
												ENST00000587575.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1700009048.hg.1 | 
												ENST00000592569.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1700009048.hg.1 | 
												ENST00000592569.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1700009048.hg.1 | 
												ENST00000592569.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1700009048.hg.1 | 
												ENST00000592569.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1700009048.hg.1 | 
												ENST00000592569.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1700009048.hg.1 | 
												NR_110845.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1700009048.hg.1 | 
												NR_110845.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1700009048.hg.1 | 
												NR_110845.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1700009048.hg.1 | 
												NR_110845.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1700009048.hg.1 | 
												NR_110845.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1700009048.hg.1 | 
												NR_110845.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1700009048.hg.1 | 
												NR_110846.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1700009048.hg.1 | 
												NR_110846.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1700009048.hg.1 | 
												NR_110846.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1700009048.hg.1 | 
												NR_110846.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1700009048.hg.1 | 
												NR_110846.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1700009048.hg.1 | 
												NR_110847.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1700009048.hg.1 | 
												NR_110847.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1700009048.hg.1 | 
												NR_110847.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1700009048.hg.1 | 
												NR_110847.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1700009048.hg.1 | 
												NR_110847.1 | 
												Refseq | 
												
											
										
										
									
								
									
										
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1700168383.hg.1 | 
												ENST00000587575.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1700168383.hg.1 | 
												ENST00000592569.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1700168383.hg.1 | 
												NR_110845.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1700168383.hg.1 | 
												NR_110846.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1700168383.hg.1 | 
												NR_110847.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1700168383.hg.1 | 
												ENST00000587575.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1700168383.hg.1 | 
												ENST00000592569.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1700168383.hg.1 | 
												NR_110845.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1700168383.hg.1 | 
												NR_110846.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1700168383.hg.1 | 
												NR_110847.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1700168383.hg.1 | 
												TC1700009048.hg.1 | 
												Clariom D TC |