Assay Details
Gene Symbol: RARA-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110861.1 | 1 - 2 | 257 | 104 | ||
GenBank mRNA | BC038432.1 | - | 1 - 2 | 257 | 104 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC105371934 | LOC105371934 | -18.4kb | Sense | Chr. 17 - 40315193 - 40322894 on NCBI Build GRCh38 | |
Downstream | gap junction protein delta 3 | GJD3 |
CX31.9, Cx30.2, GJA11, GJC1 |
17.5kb | Sense | Chr. 17 - 40360652 - 40364693 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
