Assay Details
Gene Symbol: LOC101928911 | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110869.1 | 3 - 4 | 391 | 186 | |||
GenBank mRNA | AY927641.1 | - | LIT3622 AY927641 |
3 - 4 | 392 | 186 | |
AY927642.1 | - | 2 - 3 | 240 | 186 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | akirin 2 | AKIRIN2 |
C6orf166, FBI1, dJ486L4.2 |
-207.5kb | Anti-sense | Chr. 6 - 87674860 - 87702267 on NCBI Build GRCh38 |
Downstream | sperm acrosome associated 1 | SPACA1 | SAMP32 | 136.2kb | Sense | Chr. 6 - 88046907 - 88066832 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
