Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010253.hg.1 |
ENST00000446811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010253.hg.1 |
ENST00000446811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010253.hg.1 |
ENST00000446811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010253.hg.1 |
ENST00000446811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010253.hg.1 |
ENST00000446811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010253.hg.1 |
NR_110870.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010253.hg.1 |
NR_110870.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010253.hg.1 |
NR_110870.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010253.hg.1 |
NR_110870.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010253.hg.1 |
NR_110870.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600170045.hg.1 |
ENST00000446811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600170045.hg.1 |
NR_110870.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600170045.hg.1 |
ENST00000446811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600170045.hg.1 |
NR_110870.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600170045.hg.1 |
TC0600010253.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600010253.hg.1 |
ENST00000446811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010253.hg.1 |
ENST00000446811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010253.hg.1 |
ENST00000446811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010253.hg.1 |
ENST00000446811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010253.hg.1 |
ENST00000446811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010253.hg.1 |
NR_110870.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010253.hg.1 |
NR_110870.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010253.hg.1 |
NR_110870.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010253.hg.1 |
NR_110870.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010253.hg.1 |
NR_110870.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600170044.hg.1 |
ENST00000446811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600170044.hg.1 |
NR_110870.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600170044.hg.1 |
ENST00000446811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600170044.hg.1 |
NR_110870.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600170044.hg.1 |
TC0600010253.hg.1 |
Clariom D TC |