Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000412640.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000412640.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000412640.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000412640.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000412640.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000412640.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000412640.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000412640.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000412640.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000412640.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000412640.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000412640.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000424417.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000424417.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000424417.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000424417.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000424417.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000430908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000430908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000430908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000430908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000430908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000430908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000540802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000540802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000540802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000543512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000543512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000630770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000630770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
ENST00000630770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
NR_110876.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
NR_110876.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
NR_110876.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
NR_110877.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
NR_110877.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012463.hg.1 |
NR_110877.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000412640.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000412640.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000412640.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000412640.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000412640.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000412640.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000412640.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000412640.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000412640.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000412640.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000412640.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000412640.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000424417.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000424417.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000424417.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000424417.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000424417.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000430908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000430908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000430908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000430908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000430908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000430908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000540802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000540802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000540802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000543512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000543512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000630770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000630770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
ENST00000630770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
NR_110876.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
NR_110876.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
NR_110876.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
NR_110877.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
NR_110877.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012463.hg.1 |
NR_110877.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700207648.hg.1 |
ENST00000430908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700207648.hg.1 |
ENST00000430908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700207648.hg.1 |
ENST00000543512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700207648.hg.1 |
NR_110876.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700207648.hg.1 |
ENST00000430908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700207648.hg.1 |
ENST00000430908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700207648.hg.1 |
ENST00000543512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700207648.hg.1 |
NR_110876.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700207648.hg.1 |
TC1700012463.hg.1 |
Clariom D TC |