Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012301.hg.1 |
ENST00000428865.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012301.hg.1 |
ENST00000428865.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012301.hg.1 |
ENST00000428865.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012301.hg.1 |
NR_110889.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012301.hg.1 |
NR_110889.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012301.hg.1 |
NR_110889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600184818.hg.1 |
ENST00000428865.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184818.hg.1 |
NR_110889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600184818.hg.1 |
ENST00000428865.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184818.hg.1 |
NR_110889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600184818.hg.1 |
TC0600012301.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600184819.hg.1 |
ENST00000428865.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184819.hg.1 |
NR_110889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600184819.hg.1 |
ENST00000428865.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184819.hg.1 |
NR_110889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600184819.hg.1 |
TC0600012301.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600012301.hg.1 |
ENST00000428865.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012301.hg.1 |
ENST00000428865.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012301.hg.1 |
ENST00000428865.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012301.hg.1 |
NR_110889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012301.hg.1 |
NR_110889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012301.hg.1 |
NR_110889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600184820.hg.1 |
ENST00000428865.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184820.hg.1 |
NR_110889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600184820.hg.1 |
ENST00000428865.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600184820.hg.1 |
NR_110889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600184820.hg.1 |
TC0600012301.hg.1 |
Clariom D TC |