Assay Details
| Gene Symbol: LINC01563 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110895.1 | 1 - 2 | 398 | 192 | ||
| GenBank mRNA | AB593144.1 | - | 1 - 2 | 364 | 191 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin specific peptidase 22 | USP22 | USP3L | -32.1kb | Anti-sense | Chr. 17 - 20999593 - 21043409 on NCBI Build GRCh38 |
| Downstream | dehydrogenase/reductase 7B | DHRS7B |
CGI-93, SDR32C1 |
41.4kb | Sense | Chr. 17 - 21123369 - 21191972 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LINC01563
Genomic Map