Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009466.hg.1 |
ENST00000575424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009466.hg.1 |
ENST00000575424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009466.hg.1 |
ENST00000575424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009466.hg.1 |
ENST00000576797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009466.hg.1 |
ENST00000576797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009466.hg.1 |
ENST00000576797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009466.hg.1 |
NR_110909.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009466.hg.1 |
NR_110909.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009466.hg.1 |
NR_110909.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009466.hg.1 |
ENST00000575424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009466.hg.1 |
ENST00000575424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009466.hg.1 |
ENST00000575424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009466.hg.1 |
ENST00000576797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009466.hg.1 |
ENST00000576797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009466.hg.1 |
ENST00000576797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009466.hg.1 |
NR_110909.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009466.hg.1 |
NR_110909.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009466.hg.1 |
NR_110909.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600172457.hg.1 |
ENST00000575424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600172457.hg.1 |
NR_110909.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600172457.hg.1 |
ENST00000575424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600172457.hg.1 |
NR_110909.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600172457.hg.1 |
TC1600009466.hg.1 |
Clariom D TC |