Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011018.hg.1 |
ENST00000565238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011018.hg.1 |
ENST00000565238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011018.hg.1 |
ENST00000565238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011018.hg.1 |
NR_110937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011018.hg.1 |
NR_110937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011018.hg.1 |
NR_110937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600185630.hg.1 |
ENST00000565238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600185630.hg.1 |
NR_110937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600185630.hg.1 |
ENST00000565238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600185630.hg.1 |
NR_110937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600185630.hg.1 |
TC1600011018.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600185631.hg.1 |
ENST00000565238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600185631.hg.1 |
NR_110937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600185631.hg.1 |
ENST00000565238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600185631.hg.1 |
NR_110937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600185631.hg.1 |
TC1600011018.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600011018.hg.1 |
ENST00000565238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011018.hg.1 |
ENST00000565238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011018.hg.1 |
ENST00000565238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011018.hg.1 |
NR_110937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011018.hg.1 |
NR_110937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011018.hg.1 |
NR_110937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600185632.hg.1 |
ENST00000565238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600185632.hg.1 |
NR_110937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600185632.hg.1 |
ENST00000565238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600185632.hg.1 |
NR_110937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600185632.hg.1 |
TC1600011018.hg.1 |
Clariom D TC |