Assay Details
| Gene Symbol: LOC103021296 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110984.1 | 1 - 2 | 36 | 60 | ||
| GenBank mRNA | DA449064.1 | - | 1 - 2 | 36 | 60 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 26 member 9 | SLC26A9 | Anti-sense | Chr. 1 - 205913048 - 205943472 on NCBI Build GRCh38 | ||
| Downstream | uncharacterized LOC103021295 | LOC103021295 | 22.2kb | Sense | Chr. 1 - 205957925 - 205958388 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC103021296
Genomic Map