Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0100208174.hg.1 |
ENST00000415842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208174.hg.1 |
ENST00000415842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208174.hg.1 |
ENST00000444349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208174.hg.1 |
ENST00000444349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208174.hg.1 |
NR_119383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100208174.hg.1 |
ENST00000415842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208174.hg.1 |
ENST00000415842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208174.hg.1 |
ENST00000444349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208174.hg.1 |
ENST00000444349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208174.hg.1 |
NR_119383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100208174.hg.1 |
TC0100014459.hg.1 |
Clariom D TC |