Assay Details
| Gene Symbol: LOC102724084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120307.1 | 1 - 2 | 110 | 133 | ||
| GenBank mRNA | DA228026.1 | - | 1 - 2 | 110 | 133 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1229 | LINC01229 | -727.3kb | Anti-sense | Chr. 16 - 79770471 - 79801343 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 1227 | LINC01227 | 4.0kb | Sense | Chr. 16 - 80567103 - 80572808 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000563267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000563267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000563267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000563267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000563267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000563267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000564411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000564411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000564411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000564411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000568552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000568552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000568819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000568819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000568819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000568819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000568819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000568819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000568819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | ENST00000568819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | NR_120307.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | NR_120307.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | NR_120307.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | NR_120307.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | NR_120307.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010960.hg.1 | NR_120307.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600185188.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185188.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185188.hg.1 | NR_120307.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600185188.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185188.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185188.hg.1 | NR_120307.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600185188.hg.1 | TC1600010960.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600185180.hg.1 | ENST00000563267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185180.hg.1 | ENST00000563267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185180.hg.1 | ENST00000564411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185180.hg.1 | ENST00000564411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185180.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185180.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185180.hg.1 | ENST00000568552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185180.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185180.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185180.hg.1 | ENST00000568819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185180.hg.1 | ENST00000568819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185180.hg.1 | NR_120307.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600185180.hg.1 | ENST00000563267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185180.hg.1 | ENST00000563267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185180.hg.1 | ENST00000564411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185180.hg.1 | ENST00000564411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185180.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185180.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185180.hg.1 | ENST00000568552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185180.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185180.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185180.hg.1 | ENST00000568819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185180.hg.1 | ENST00000568819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185180.hg.1 | NR_120307.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600185180.hg.1 | TC1600010960.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000563267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000563267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000563267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000563267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000563267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000563267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000564411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000564411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000564411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000564411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000565050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000568552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000568552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000568776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000568819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000568819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000568819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000568819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000568819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000568819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000568819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | ENST00000568819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | NR_120307.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | NR_120307.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | NR_120307.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | NR_120307.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | NR_120307.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010960.hg.1 | NR_120307.1 | Refseq |

LOC102724084
Genomic Map