Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010902.hg.1 |
ENST00000560778.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010902.hg.1 |
ENST00000560778.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010902.hg.1 |
ENST00000560778.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010902.hg.1 |
ENST00000560778.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010902.hg.1 |
ENST00000560778.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010902.hg.1 |
ENST00000560778.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010902.hg.1 |
ENST00000560778.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010902.hg.1 |
NR_120317.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010902.hg.1 |
NR_120317.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010902.hg.1 |
NR_120317.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010902.hg.1 |
NR_120317.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010902.hg.1 |
NR_120317.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010902.hg.1 |
NR_120317.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010902.hg.1 |
NR_120317.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010902.hg.1 |
NR_120317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185617.hg.1 |
ENST00000560778.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185617.hg.1 |
NR_120317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185617.hg.1 |
ENST00000560778.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185617.hg.1 |
NR_120317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185617.hg.1 |
TC1500010902.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500185616.hg.1 |
ENST00000560778.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185616.hg.1 |
NR_120317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185616.hg.1 |
ENST00000560778.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185616.hg.1 |
NR_120317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185616.hg.1 |
TC1500010902.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500185619.hg.1 |
NR_120317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185619.hg.1 |
NR_120317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185619.hg.1 |
TC1500010902.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500010902.hg.1 |
ENST00000560778.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010902.hg.1 |
ENST00000560778.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010902.hg.1 |
ENST00000560778.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010902.hg.1 |
ENST00000560778.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010902.hg.1 |
ENST00000560778.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010902.hg.1 |
ENST00000560778.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010902.hg.1 |
ENST00000560778.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010902.hg.1 |
NR_120317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010902.hg.1 |
NR_120317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010902.hg.1 |
NR_120317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010902.hg.1 |
NR_120317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010902.hg.1 |
NR_120317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010902.hg.1 |
NR_120317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010902.hg.1 |
NR_120317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010902.hg.1 |
NR_120317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185618.hg.1 |
ENST00000560778.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185618.hg.1 |
NR_120317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185618.hg.1 |
ENST00000560778.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185618.hg.1 |
NR_120317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185618.hg.1 |
TC1500010902.hg.1 |
Clariom D TC |