Assay Details
Gene Symbol: LOC101928174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120329.1 | 2 - 3 | 297 | 76 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | gap junction protein delta 2 | GJD2 |
CX36, GJA9 |
-22.7kb | Anti-sense | Chr. 15 - 34751746 - 34754955 on NCBI Build GRCh38 |
Downstream | aquarius intron-binding spliceosomal factor | AQR |
IBP160, fSAP164 |
43.4kb | Anti-sense | Chr. 15 - 34856351 - 34969794 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1800006710.hg.1 | ENST00000526991.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006710.hg.1 | ENST00000526991.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006710.hg.1 | ENST00000526991.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006710.hg.1 | ENST00000526991.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006710.hg.1 | ENST00000526991.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006710.hg.1 | ENST00000526991.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006710.hg.1 | ENST00000526991.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006710.hg.1 | ENST00000526991.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006710.hg.1 | ENST00000526991.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006710.hg.1 | ENST00000526991.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006710.hg.1 | ENST00000588284.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006710.hg.1 | ENST00000588284.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006710.hg.1 | NM_020412.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006710.hg.1 | NM_020412.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006710.hg.1 | NM_020412.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006710.hg.1 | NM_020412.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006710.hg.1 | NM_020412.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006710.hg.1 | NM_020412.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006710.hg.1 | NM_020412.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006710.hg.1 | NM_020412.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006710.hg.1 | NM_020412.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006710.hg.1 | NM_020412.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006836.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006836.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006836.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006836.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006836.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006836.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006836.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006836.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006836.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006836.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006836.hg.1 | ENST00000558707.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006836.hg.1 | ENST00000558707.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006836.hg.1 | ENST00000558707.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006836.hg.1 | NR_120329.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006836.hg.1 | NR_120329.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006836.hg.1 | NR_120329.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006836.hg.1 | NR_120329.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006836.hg.1 | NR_120329.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006836.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006836.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006836.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006836.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006836.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006836.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006836.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006836.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006836.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006836.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006836.hg.1 | ENST00000558707.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006836.hg.1 | ENST00000558707.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006836.hg.1 | ENST00000558707.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006836.hg.1 | NR_120329.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006836.hg.1 | NR_120329.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006836.hg.1 | NR_120329.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006836.hg.1 | NR_120329.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006836.hg.1 | NR_120329.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500147515.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500147515.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500147515.hg.1 | NR_120329.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500147515.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500147515.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500147515.hg.1 | NR_120329.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500147515.hg.1 | TC1500006836.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1800006710.hg.1 | ENST00000526991.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006710.hg.1 | ENST00000526991.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006710.hg.1 | ENST00000526991.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006710.hg.1 | ENST00000526991.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006710.hg.1 | ENST00000526991.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006710.hg.1 | ENST00000526991.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006710.hg.1 | ENST00000526991.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006710.hg.1 | ENST00000526991.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006710.hg.1 | ENST00000526991.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006710.hg.1 | ENST00000526991.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006710.hg.1 | ENST00000588284.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006710.hg.1 | ENST00000588284.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006710.hg.1 | NM_020412.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006710.hg.1 | NM_020412.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006710.hg.1 | NM_020412.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006710.hg.1 | NM_020412.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006710.hg.1 | NM_020412.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006710.hg.1 | NM_020412.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006710.hg.1 | NM_020412.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006710.hg.1 | NM_020412.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006710.hg.1 | NM_020412.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006710.hg.1 | NM_020412.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500147514.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500147514.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500147514.hg.1 | NR_120329.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500147514.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500147514.hg.1 | ENST00000503496.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500147514.hg.1 | NR_120329.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500147514.hg.1 | TC1500006836.hg.1 | Clariom D TC |
