Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009330.hg.1 |
ENST00000558434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009330.hg.1 |
ENST00000558434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009330.hg.1 |
ENST00000558434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009330.hg.1 |
ENST00000558434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009330.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009330.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009330.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009330.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009330.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009330.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009330.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009330.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009330.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009330.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009330.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009330.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009330.hg.1 |
ENST00000561238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009330.hg.1 |
ENST00000561238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009330.hg.1 |
ENST00000561238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009330.hg.1 |
ENST00000561238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009330.hg.1 |
ENST00000561238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009330.hg.1 |
NR_120336.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009330.hg.1 |
NR_120336.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009330.hg.1 |
NR_120336.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009330.hg.1 |
NR_120336.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009330.hg.1 |
NR_120336.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009330.hg.1 |
NR_120336.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009330.hg.1 |
ENST00000558434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009330.hg.1 |
ENST00000558434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009330.hg.1 |
ENST00000558434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009330.hg.1 |
ENST00000558434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009330.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009330.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009330.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009330.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009330.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009330.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009330.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009330.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009330.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009330.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009330.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009330.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009330.hg.1 |
ENST00000561238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009330.hg.1 |
ENST00000561238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009330.hg.1 |
ENST00000561238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009330.hg.1 |
ENST00000561238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009330.hg.1 |
ENST00000561238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009330.hg.1 |
NR_120336.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009330.hg.1 |
NR_120336.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009330.hg.1 |
NR_120336.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009330.hg.1 |
NR_120336.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009330.hg.1 |
NR_120336.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009330.hg.1 |
NR_120336.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500168350.hg.1 |
ENST00000558434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168350.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168350.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168350.hg.1 |
ENST00000561238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168350.hg.1 |
NR_120336.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500168350.hg.1 |
ENST00000558434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168350.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168350.hg.1 |
ENST00000558792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168350.hg.1 |
ENST00000561238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168350.hg.1 |
NR_120336.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500168350.hg.1 |
TC1500009330.hg.1 |
Clariom D TC |