Assay Details
Gene Symbol: LOC102723344 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120375.1 | 1 - 2 | 30 | 78 | ||
GenBank mRNA | BX642193.1 | - | 1 - 2 | 125 | 78 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | carbonic anhydrase 12 | CA12 |
CA-XII, CAXII, HsT18816, T18816 |
-8.1kb | Anti-sense | Chr. 15 - 63321378 - 63382166 on NCBI Build GRCh38 |
Downstream | ubiquitin specific peptidase 3 | USP3 |
SIH003, UBP |
109.6kb | Sense | Chr. 15 - 63504511 - 63594640 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500153309.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153309.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153309.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153309.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153309.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500153309.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153309.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153309.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153309.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153309.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500153309.hg.1 | TC1500007524.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500153308.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153308.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153308.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153308.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153308.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500153308.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153308.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153308.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153308.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153308.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500153308.hg.1 | TC1500007524.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500153307.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153307.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153307.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153307.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153307.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500153307.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153307.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153307.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153307.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153307.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500153307.hg.1 | TC1500007524.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500153312.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153312.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153312.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153312.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153312.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500153312.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153312.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153312.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153312.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153312.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500153312.hg.1 | TC1500007524.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500153306.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153306.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153306.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153306.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153306.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500153306.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153306.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153306.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153306.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500153306.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500153306.hg.1 | TC1500007524.hg.1 | Clariom D TC |
