| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008145.hg.1 |
ENST00000559866.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008145.hg.1 |
ENST00000559866.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008145.hg.1 |
ENST00000559866.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008145.hg.1 |
ENST00000559866.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008145.hg.1 |
ENST00000559866.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008145.hg.1 |
NR_120379.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008145.hg.1 |
NR_120379.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008145.hg.1 |
NR_120379.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008145.hg.1 |
NR_120379.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008145.hg.1 |
NR_120379.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008145.hg.1 |
NR_120379.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008092.hg.1 |
ENST00000456932.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008092.hg.1 |
ENST00000456932.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008092.hg.1 |
ENST00000456932.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008092.hg.1 |
ENST00000456932.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008092.hg.1 |
ENST00000456932.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008092.hg.1 |
ENST00000456932.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008092.hg.1 |
ENST00000456932.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008092.hg.1 |
ENST00000456932.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008092.hg.1 |
ENST00000612093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008092.hg.1 |
ENST00000612093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008092.hg.1 |
ENST00000612093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008092.hg.1 |
ENST00000612093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008092.hg.1 |
ENST00000612093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008092.hg.1 |
ENST00000612093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008092.hg.1 |
ENST00000612093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008092.hg.1 |
ENST00000612093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008092.hg.1 |
ENST00000612093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008092.hg.1 |
ENST00000612093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008092.hg.1 |
ENST00000612093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008092.hg.1 |
ENST00000612093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008092.hg.1 |
ENST00000617986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500158597.hg.1 |
ENST00000559866.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500158597.hg.1 |
NR_120379.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500158597.hg.1 |
ENST00000559866.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500158597.hg.1 |
NR_120379.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500158597.hg.1 |
TC1500008145.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500158598.hg.1 |
ENST00000559866.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500158598.hg.1 |
NR_120379.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500158598.hg.1 |
ENST00000559866.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500158598.hg.1 |
NR_120379.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500158598.hg.1 |
TC1500008145.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500158595.hg.1 |
ENST00000559866.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500158595.hg.1 |
NR_120379.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500158595.hg.1 |
ENST00000559866.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500158595.hg.1 |
NR_120379.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500158595.hg.1 |
TC1500008145.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500158599.hg.1 |
ENST00000559866.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500158599.hg.1 |
NR_120379.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500158599.hg.1 |
ENST00000559866.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500158599.hg.1 |
NR_120379.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500158599.hg.1 |
TC1500008145.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500158596.hg.1 |
ENST00000559866.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500158596.hg.1 |
NR_120379.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500158596.hg.1 |
ENST00000559866.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500158596.hg.1 |
NR_120379.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500158596.hg.1 |
TC1500008145.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1500008145.hg.1 |
ENST00000559866.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008145.hg.1 |
ENST00000559866.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008145.hg.1 |
ENST00000559866.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008145.hg.1 |
ENST00000559866.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008145.hg.1 |
ENST00000559866.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008145.hg.1 |
NR_120379.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008145.hg.1 |
NR_120379.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008145.hg.1 |
NR_120379.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008145.hg.1 |
NR_120379.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008145.hg.1 |
NR_120379.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008145.hg.1 |
NR_120379.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500158594.hg.1 |
NR_120379.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500158594.hg.1 |
NR_120379.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500158594.hg.1 |
TC1500008145.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1500008092.hg.1 |
ENST00000456932.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008092.hg.1 |
ENST00000456932.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008092.hg.1 |
ENST00000456932.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008092.hg.1 |
ENST00000456932.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008092.hg.1 |
ENST00000456932.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008092.hg.1 |
ENST00000456932.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008092.hg.1 |
ENST00000456932.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008092.hg.1 |
ENST00000456932.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008092.hg.1 |
ENST00000612093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008092.hg.1 |
ENST00000612093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008092.hg.1 |
ENST00000612093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008092.hg.1 |
ENST00000612093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008092.hg.1 |
ENST00000612093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008092.hg.1 |
ENST00000612093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008092.hg.1 |
ENST00000612093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008092.hg.1 |
ENST00000612093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008092.hg.1 |
ENST00000612093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008092.hg.1 |
ENST00000612093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008092.hg.1 |
ENST00000612093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008092.hg.1 |
ENST00000612093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008092.hg.1 |
ENST00000617986.1 |
Ensembl |