Assay Details
| Gene Symbol: PCSK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120409.1 | 9 - 10 | 1745 | 69 | ||
| XM_017014800.1 | XP_016870289.1 | 2 - 3 | 165 | 69 | ||
| GenBank mRNA | AK225773.1 | - | 9 - 10 | 1742 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105376090 | LOC105376090 | -926.6kb | Sense | Chr. 9 - 75151611 - 75179773 on NCBI Build GRCh38 | |
| Downstream | riboflavin kinase | RFK | RIFK | 228.4kb | Anti-sense | Chr. 9 - 76385517 - 76394528 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017014800.1 transcripts.
Set Membership: |
|

PCSK5
Genomic Map