Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007315.hg.1 |
ENST00000615202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007315.hg.1 |
ENST00000615202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007315.hg.1 |
ENST00000615202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007315.hg.1 |
NR_120416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007315.hg.1 |
NR_120416.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007315.hg.1 |
NR_120416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149581.hg.1 |
ENST00000615202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149581.hg.1 |
NR_120416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149581.hg.1 |
ENST00000615202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149581.hg.1 |
NR_120416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149581.hg.1 |
TC1300007315.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300149582.hg.1 |
ENST00000615202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149582.hg.1 |
NR_120416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149582.hg.1 |
ENST00000615202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149582.hg.1 |
NR_120416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149582.hg.1 |
TC1300007315.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300007315.hg.1 |
ENST00000615202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007315.hg.1 |
ENST00000615202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007315.hg.1 |
ENST00000615202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007315.hg.1 |
NR_120416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007315.hg.1 |
NR_120416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007315.hg.1 |
NR_120416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149580.hg.1 |
ENST00000615202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149580.hg.1 |
NR_120416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149580.hg.1 |
ENST00000615202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149580.hg.1 |
NR_120416.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149580.hg.1 |
TC1300007315.hg.1 |
Clariom D TC |