Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000203.hg.1 |
ENST00000299183.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000203.hg.1 |
ENST00000347735.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000203.hg.1 |
ENST00000355446.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000203.hg.1 |
ENST00000360905.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000203.hg.1 |
ENST00000397203.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000203.hg.1 |
ENST00000440394.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000203.hg.1 |
ENST00000454194.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000203.hg.1 |
ENST00000536573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000203.hg.1 |
ENST00000537890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000203.hg.1 |
ENST00000538971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000203.hg.1 |
ENST00000542302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000203.hg.1 |
ENST00000543086.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000203.hg.1 |
ENST00000545948.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000203.hg.1 |
ENST00000546231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000203.hg.1 |
NM_178039.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000203.hg.1 |
NM_178040.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000203.hg.1 |
NR_027946.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000203.hg.1 |
NR_027948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000203.hg.1 |
NR_027949.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000203.hg.1 |
TC12000014.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000347735.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000355446.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000360905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000397203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000440394.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000536573.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000538971.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000541503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000542302.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000543086.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000545948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000546231.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000589028.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000611180.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
NM_001301248.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
NM_178039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
NM_178040.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
NR_027946.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
NR_027948.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
NR_027949.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000347735.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000355446.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000360905.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000397203.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000440394.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000536573.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000538971.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000541503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000542302.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000543086.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000545948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000546231.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000589028.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
ENST00000611180.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
NM_001301248.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
NM_178039.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
NM_178040.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
NR_027946.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
NR_027948.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
NR_027949.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200144346.hg.1 |
TC1200006457.hg.1 |
Clariom D TC |