Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012822.hg.1 |
ENST00000360485.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012822.hg.1 |
ENST00000360485.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012822.hg.1 |
ENST00000360485.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012822.hg.1 |
ENST00000360485.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012822.hg.1 |
ENST00000536112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012822.hg.1 |
ENST00000536112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012822.hg.1 |
ENST00000536112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012822.hg.1 |
ENST00000536112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012822.hg.1 |
ENST00000542875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012822.hg.1 |
ENST00000542875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012822.hg.1 |
ENST00000542875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012822.hg.1 |
ENST00000542875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012822.hg.1 |
ENST00000542875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012822.hg.1 |
NR_120458.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012822.hg.1 |
NR_120458.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012822.hg.1 |
NR_120458.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012822.hg.1 |
NR_120458.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012822.hg.1 |
NR_120458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200204065.hg.1 |
ENST00000360485.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204065.hg.1 |
ENST00000536112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204065.hg.1 |
NR_120458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200204065.hg.1 |
ENST00000360485.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204065.hg.1 |
ENST00000536112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204065.hg.1 |
NR_120458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200204065.hg.1 |
TC1200012822.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200204066.hg.1 |
ENST00000360485.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204066.hg.1 |
ENST00000536112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204066.hg.1 |
NR_120458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200204066.hg.1 |
ENST00000360485.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204066.hg.1 |
ENST00000536112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204066.hg.1 |
NR_120458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200204066.hg.1 |
TC1200012822.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200204071.hg.1 |
ENST00000360485.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204071.hg.1 |
NR_120458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200204071.hg.1 |
ENST00000360485.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204071.hg.1 |
NR_120458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200204071.hg.1 |
TC1200012822.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200204079.hg.1 |
ENST00000360485.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204079.hg.1 |
ENST00000542875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204079.hg.1 |
NR_120458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200204079.hg.1 |
ENST00000360485.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204079.hg.1 |
ENST00000542875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204079.hg.1 |
NR_120458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200204079.hg.1 |
TC1200012822.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200204082.hg.1 |
ENST00000542875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204082.hg.1 |
NR_120458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200204082.hg.1 |
ENST00000542875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204082.hg.1 |
NR_120458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200204082.hg.1 |
TC1200012822.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200012822.hg.1 |
ENST00000360485.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012822.hg.1 |
ENST00000360485.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012822.hg.1 |
ENST00000360485.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012822.hg.1 |
ENST00000360485.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012822.hg.1 |
ENST00000536112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012822.hg.1 |
ENST00000536112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012822.hg.1 |
ENST00000536112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012822.hg.1 |
ENST00000536112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012822.hg.1 |
ENST00000542875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012822.hg.1 |
ENST00000542875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012822.hg.1 |
ENST00000542875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012822.hg.1 |
ENST00000542875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012822.hg.1 |
ENST00000542875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012822.hg.1 |
NR_120458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012822.hg.1 |
NR_120458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012822.hg.1 |
NR_120458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012822.hg.1 |
NR_120458.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012822.hg.1 |
NR_120458.1 |
Refseq |