Assay Details
| Gene Symbol: LINC01481 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120460.1 | 1 - 2 | 51 | 65 | ||
| GenBank mRNA | DA349921.1 | - | 1 - 2 | 49 | 65 | |
| HY013176.1 | - | 1 - 2 | 51 | 65 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | myelin regulatory factor-like | MYRFL |
C12orf15, C12orf28, bcm1377 |
-284.1kb | Anti-sense | Chr. 12 - 69824518 - 69958726 on NCBI Build GRCh38 |
| Downstream | potassium calcium-activated channel subfamily M regulatory beta subunit 4 | KCNMB4 | 122.9kb | Anti-sense | Chr. 12 - 70366220 - 70434292 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000552998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000552998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000553135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000553135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000553135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000553135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187068.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187068.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187068.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187068.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187068.hg.1 | ENST00000553135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187068.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187068.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187068.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187068.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187068.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187068.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187068.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187068.hg.1 | ENST00000553135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187068.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187068.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187068.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187068.hg.1 | TC1200011230.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000552998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000552998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000553135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000553135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000553135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000553135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187065.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187065.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187065.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187065.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187065.hg.1 | ENST00000553135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187065.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187065.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187065.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187065.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187065.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187065.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187065.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187065.hg.1 | ENST00000553135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187065.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187065.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187065.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187065.hg.1 | TC1200011230.hg.1 | Clariom D TC |

LINC01481
Genomic Map