Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006942.hg.1 |
ENST00000542401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006942.hg.1 |
ENST00000542401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006942.hg.1 |
ENST00000542401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006942.hg.1 |
ENST00000542401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006942.hg.1 |
ENST00000542401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006942.hg.1 |
ENST00000542401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006942.hg.1 |
ENST00000545424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006942.hg.1 |
ENST00000545424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006942.hg.1 |
ENST00000545424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006942.hg.1 |
ENST00000545424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006942.hg.1 |
ENST00000545424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006942.hg.1 |
ENST00000545424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006942.hg.1 |
ENST00000545424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006942.hg.1 |
ENST00000545424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006942.hg.1 |
ENST00000545424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006942.hg.1 |
ENST00000545424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006942.hg.1 |
NR_120465.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006942.hg.1 |
NR_120465.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006942.hg.1 |
NR_120465.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006942.hg.1 |
NR_120465.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006942.hg.1 |
NR_120465.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006942.hg.1 |
NR_120465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148525.hg.1 |
ENST00000542401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148525.hg.1 |
ENST00000545424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148525.hg.1 |
ENST00000545424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148525.hg.1 |
NR_120465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148525.hg.1 |
ENST00000542401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148525.hg.1 |
ENST00000545424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148525.hg.1 |
ENST00000545424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148525.hg.1 |
NR_120465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148525.hg.1 |
TC1200006942.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200006942.hg.1 |
ENST00000542401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006942.hg.1 |
ENST00000542401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006942.hg.1 |
ENST00000542401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006942.hg.1 |
ENST00000542401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006942.hg.1 |
ENST00000542401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006942.hg.1 |
ENST00000542401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006942.hg.1 |
ENST00000545424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006942.hg.1 |
ENST00000545424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006942.hg.1 |
ENST00000545424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006942.hg.1 |
ENST00000545424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006942.hg.1 |
ENST00000545424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006942.hg.1 |
ENST00000545424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006942.hg.1 |
ENST00000545424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006942.hg.1 |
ENST00000545424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006942.hg.1 |
ENST00000545424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006942.hg.1 |
ENST00000545424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006942.hg.1 |
NR_120465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006942.hg.1 |
NR_120465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006942.hg.1 |
NR_120465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006942.hg.1 |
NR_120465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006942.hg.1 |
NR_120465.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006942.hg.1 |
NR_120465.1 |
Refseq |