Assay Details
Gene Symbol: CALML3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120497.1 | 1 - 2 | 113 | 75 | ||
GenBank mRNA | DA220455.1 | - | 1 - 2 | 115 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | calmodulin like 5 | CALML5 | CLSP | -22.3kb | Sense | Chr. 10 - 5498695 - 5499570 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC105376382 | LOC105376382 | 68.7kb | Sense | Chr. 10 - 5594984 - 5596118 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000545372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000545372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000545372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000545372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166037.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166037.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166037.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166037.hg.1 | ENST00000545372.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166037.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166037.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166037.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166037.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166037.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166037.hg.1 | ENST00000545372.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166037.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166037.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166037.hg.1 | TC1000009667.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000545372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000545372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000545372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000545372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166040.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166040.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166040.hg.1 | ENST00000545372.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166040.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166040.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166040.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166040.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166040.hg.1 | ENST00000545372.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166040.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166040.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166040.hg.1 | TC1000009667.hg.1 | Clariom D TC |
