Assay Details
Gene Symbol: LINC01510 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120506.1 | 3 - 4 | 671 | 91 | ||
GenBank mRNA | CR745839.1 | - | 2 - 3 | 421 | 91 | |
LN812954.1 | - | 3 - 4 | 636 | 91 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | caveolin 1 | CAV1 |
BSCL3, CGL3, LCCNS, MSTP085, PPH3, VIP21 |
-9.9kb | Anti-sense | Chr. 7 - 116524785 - 116561185 on NCBI Build GRCh38 |
Downstream |
MET proto-oncogene, receptor tyrosine kinase |
MET |
AUTS9, DFNB97, HGFR, RCCP2, c-Met |
98.7kb | Anti-sense | Chr. 7 - 116672359 - 116798386 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000421965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000421965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000421965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000421965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000421965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000421965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000441991.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000441991.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000441991.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000441991.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000441991.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000450063.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000450063.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000450063.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000450063.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700189013.hg.1 | ENST00000441991.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189013.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189013.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700189013.hg.1 | ENST00000441991.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189013.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189013.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700189013.hg.1 | TC0700012344.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700189014.hg.1 | ENST00000441991.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189014.hg.1 | ENST00000450063.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189014.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189014.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700189014.hg.1 | ENST00000441991.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189014.hg.1 | ENST00000450063.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189014.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189014.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700189014.hg.1 | TC0700012344.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700189011.hg.1 | ENST00000441991.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189011.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700189011.hg.1 | ENST00000441991.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189011.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700189011.hg.1 | TC0700012344.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700189017.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189017.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700189017.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189017.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700189017.hg.1 | TC0700012344.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700189018.hg.1 | ENST00000441991.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189018.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189018.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700189018.hg.1 | ENST00000441991.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189018.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189018.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700189018.hg.1 | TC0700012344.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700189021.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189021.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700189021.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189021.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700189021.hg.1 | TC0700012344.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700189022.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189022.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700189022.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189022.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700189022.hg.1 | TC0700012344.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000421965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000421965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000421965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000421965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000421965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000421965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000441991.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000441991.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000441991.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000441991.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000441991.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000450063.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000450063.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000450063.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000450063.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700189009.hg.1 | ENST00000421965.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189009.hg.1 | ENST00000441991.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189009.hg.1 | ENST00000450063.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189009.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700189009.hg.1 | ENST00000421965.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189009.hg.1 | ENST00000441991.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189009.hg.1 | ENST00000450063.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189009.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700189009.hg.1 | TC0700012344.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700189016.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189016.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700189016.hg.1 | ENST00000458082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189016.hg.1 | NR_120506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700189016.hg.1 | TC0700012344.hg.1 | Clariom D TC |
