Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
ENST00000421965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
ENST00000421965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
ENST00000421965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
ENST00000421965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
ENST00000421965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
ENST00000421965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
ENST00000441991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
ENST00000441991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
ENST00000441991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
ENST00000441991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
ENST00000441991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
ENST00000450063.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
ENST00000450063.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
ENST00000450063.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
ENST00000450063.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
ENST00000458082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
ENST00000458082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
ENST00000458082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
ENST00000458082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
ENST00000458082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
ENST00000458082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
ENST00000458082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
ENST00000421965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
ENST00000421965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
ENST00000421965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
ENST00000421965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
ENST00000421965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
ENST00000421965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
ENST00000441991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
ENST00000441991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
ENST00000441991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
ENST00000441991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
ENST00000441991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
ENST00000450063.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
ENST00000450063.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
ENST00000450063.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
ENST00000450063.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
ENST00000458082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
ENST00000458082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
ENST00000458082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
ENST00000458082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
ENST00000458082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
ENST00000458082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
ENST00000458082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012344.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700189000.hg.1 |
ENST00000421965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189000.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700189000.hg.1 |
ENST00000421965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189000.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700189000.hg.1 |
TC0700012344.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700189001.hg.1 |
ENST00000421965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189001.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700189001.hg.1 |
ENST00000421965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189001.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700189001.hg.1 |
TC0700012344.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700189002.hg.1 |
ENST00000421965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189002.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700189002.hg.1 |
ENST00000421965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189002.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700189002.hg.1 |
TC0700012344.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700189003.hg.1 |
ENST00000421965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189003.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700189003.hg.1 |
ENST00000421965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189003.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700189003.hg.1 |
TC0700012344.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700189008.hg.1 |
ENST00000421965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189008.hg.1 |
ENST00000450063.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189008.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700189008.hg.1 |
ENST00000421965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189008.hg.1 |
ENST00000450063.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189008.hg.1 |
NR_120506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700189008.hg.1 |
TC0700012344.hg.1 |
Clariom D TC |