Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011460.hg.1 |
ENST00000511013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011460.hg.1 |
ENST00000511013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011460.hg.1 |
ENST00000511013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011460.hg.1 |
ENST00000528316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011460.hg.1 |
ENST00000528316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011460.hg.1 |
ENST00000528316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011460.hg.1 |
ENST00000528316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011460.hg.1 |
NR_120529.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011460.hg.1 |
NR_120529.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011460.hg.1 |
NR_120529.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011460.hg.1 |
NR_120530.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011460.hg.1 |
NR_120530.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011460.hg.1 |
ENST00000511013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011460.hg.1 |
ENST00000511013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011460.hg.1 |
ENST00000511013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011460.hg.1 |
ENST00000528316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011460.hg.1 |
ENST00000528316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011460.hg.1 |
ENST00000528316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011460.hg.1 |
ENST00000528316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011460.hg.1 |
NR_120529.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011460.hg.1 |
NR_120529.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011460.hg.1 |
NR_120529.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011460.hg.1 |
NR_120530.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011460.hg.1 |
NR_120530.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188333.hg.1 |
ENST00000511013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188333.hg.1 |
NR_120529.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188333.hg.1 |
ENST00000511013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188333.hg.1 |
NR_120529.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188333.hg.1 |
TC1100011460.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100188334.hg.1 |
ENST00000511013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188334.hg.1 |
NR_120529.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188334.hg.1 |
ENST00000511013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188334.hg.1 |
NR_120529.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188334.hg.1 |
TC1100011460.hg.1 |
Clariom D TC |