Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007237.hg.1 |
ENST00000530352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007237.hg.1 |
ENST00000530352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007237.hg.1 |
ENST00000530352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007237.hg.1 |
ENST00000533046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007237.hg.1 |
ENST00000533046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007237.hg.1 |
ENST00000533046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007237.hg.1 |
ENST00000533046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007237.hg.1 |
ENST00000533046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007237.hg.1 |
ENST00000533046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007237.hg.1 |
NR_120550.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007237.hg.1 |
NR_120550.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007237.hg.1 |
NR_120550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150973.hg.1 |
ENST00000530352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150973.hg.1 |
ENST00000533046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150973.hg.1 |
ENST00000533046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150973.hg.1 |
NR_120550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150973.hg.1 |
ENST00000530352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150973.hg.1 |
ENST00000533046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150973.hg.1 |
ENST00000533046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150973.hg.1 |
NR_120550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150973.hg.1 |
TC1100007237.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007237.hg.1 |
ENST00000530352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007237.hg.1 |
ENST00000530352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007237.hg.1 |
ENST00000530352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007237.hg.1 |
ENST00000533046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007237.hg.1 |
ENST00000533046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007237.hg.1 |
ENST00000533046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007237.hg.1 |
ENST00000533046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007237.hg.1 |
ENST00000533046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007237.hg.1 |
ENST00000533046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007237.hg.1 |
NR_120550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007237.hg.1 |
NR_120550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007237.hg.1 |
NR_120550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150975.hg.1 |
ENST00000530352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150975.hg.1 |
ENST00000533046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150975.hg.1 |
ENST00000533046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150975.hg.1 |
NR_120550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150975.hg.1 |
ENST00000530352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150975.hg.1 |
ENST00000533046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150975.hg.1 |
ENST00000533046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150975.hg.1 |
NR_120550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150975.hg.1 |
TC1100007237.hg.1 |
Clariom D TC |