Assay Details
| Gene Symbol: LINC01493 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120553.1 | 2 - 3 | 214 | 98 | ||
| GenBank mRNA | AI218577.1 | - | 1 - 2 | 98 | ||
| BX099902.1 | - | 2 - 3 | 98 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC103312105 | LOC103312105 | -6.6kb | Anti-sense | Chr. 11 - 38618264 - 38646356 on NCBI Build GRCh38 | |
| Downstream | leucine rich repeat containing 4C | LRRC4C |
NGL-1, NGL1 |
1.5mb | Anti-sense | Chr. 11 - 40107066 - 41460212 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC01493
Genomic Map