Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011697.hg.1 |
ENST00000526585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011697.hg.1 |
ENST00000526585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011697.hg.1 |
ENST00000526585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011697.hg.1 |
ENST00000527778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011697.hg.1 |
ENST00000527778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011697.hg.1 |
ENST00000527778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011697.hg.1 |
ENST00000528075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011697.hg.1 |
ENST00000528075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011697.hg.1 |
ENST00000528075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011697.hg.1 |
ENST00000528075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011697.hg.1 |
NR_120561.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011697.hg.1 |
NR_120561.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011697.hg.1 |
NR_120561.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011697.hg.1 |
NR_120561.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011697.hg.1 |
NR_120562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011697.hg.1 |
NR_120562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011697.hg.1 |
NR_120562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011697.hg.1 |
NR_120562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100190279.hg.1 |
ENST00000526585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100190279.hg.1 |
ENST00000527778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100190279.hg.1 |
ENST00000528075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100190279.hg.1 |
NR_120561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100190279.hg.1 |
NR_120562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100190279.hg.1 |
ENST00000526585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100190279.hg.1 |
ENST00000527778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100190279.hg.1 |
ENST00000528075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100190279.hg.1 |
NR_120561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100190279.hg.1 |
NR_120562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100190279.hg.1 |
TC1100011697.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100011697.hg.1 |
ENST00000526585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011697.hg.1 |
ENST00000526585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011697.hg.1 |
ENST00000526585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011697.hg.1 |
ENST00000527778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011697.hg.1 |
ENST00000527778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011697.hg.1 |
ENST00000527778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011697.hg.1 |
ENST00000528075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011697.hg.1 |
ENST00000528075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011697.hg.1 |
ENST00000528075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011697.hg.1 |
ENST00000528075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011697.hg.1 |
NR_120561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011697.hg.1 |
NR_120561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011697.hg.1 |
NR_120561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011697.hg.1 |
NR_120561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011697.hg.1 |
NR_120562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011697.hg.1 |
NR_120562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011697.hg.1 |
NR_120562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011697.hg.1 |
NR_120562.1 |
Refseq |