Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010731.hg.1 |
ENST00000543925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010731.hg.1 |
ENST00000543925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010731.hg.1 |
ENST00000543925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010731.hg.1 |
ENST00000543925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010731.hg.1 |
ENST00000543925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010731.hg.1 |
ENST00000543925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010731.hg.1 |
ENST00000545474.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010731.hg.1 |
ENST00000545474.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010731.hg.1 |
ENST00000545474.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010731.hg.1 |
NR_120568.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010731.hg.1 |
NR_120568.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010731.hg.1 |
NR_120568.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010731.hg.1 |
NR_120569.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010731.hg.1 |
NR_120569.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010731.hg.1 |
NR_120569.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010731.hg.1 |
ENST00000543925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010731.hg.1 |
ENST00000543925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010731.hg.1 |
ENST00000543925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010731.hg.1 |
ENST00000543925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010731.hg.1 |
ENST00000543925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010731.hg.1 |
ENST00000543925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010731.hg.1 |
ENST00000545474.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010731.hg.1 |
ENST00000545474.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010731.hg.1 |
ENST00000545474.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010731.hg.1 |
NR_120568.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010731.hg.1 |
NR_120568.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010731.hg.1 |
NR_120568.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010731.hg.1 |
NR_120569.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010731.hg.1 |
NR_120569.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010731.hg.1 |
NR_120569.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100181292.hg.1 |
ENST00000543925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181292.hg.1 |
ENST00000543925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181292.hg.1 |
ENST00000545474.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181292.hg.1 |
NR_120568.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100181292.hg.1 |
NR_120569.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100181292.hg.1 |
ENST00000543925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181292.hg.1 |
ENST00000543925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181292.hg.1 |
ENST00000545474.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181292.hg.1 |
NR_120568.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100181292.hg.1 |
NR_120569.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100181292.hg.1 |
TC1100010731.hg.1 |
Clariom D TC |