Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009400.hg.1 |
ENST00000504932.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009400.hg.1 |
ENST00000504932.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009400.hg.1 |
ENST00000504932.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009400.hg.1 |
ENST00000504932.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009400.hg.1 |
ENST00000532579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009400.hg.1 |
ENST00000532579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009400.hg.1 |
ENST00000532579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009400.hg.1 |
NR_120577.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009400.hg.1 |
NR_120577.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009400.hg.1 |
NR_120577.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009400.hg.1 |
NR_120577.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009400.hg.1 |
NR_120579.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009400.hg.1 |
NR_120579.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009400.hg.1 |
NR_120579.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009400.hg.1 |
NR_120579.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170402.hg.1 |
ENST00000504932.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170402.hg.1 |
NR_120577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170402.hg.1 |
ENST00000504932.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170402.hg.1 |
NR_120577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170402.hg.1 |
TC1100009400.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100009400.hg.1 |
ENST00000504932.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009400.hg.1 |
ENST00000504932.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009400.hg.1 |
ENST00000504932.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009400.hg.1 |
ENST00000504932.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009400.hg.1 |
ENST00000532579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009400.hg.1 |
ENST00000532579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009400.hg.1 |
ENST00000532579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009400.hg.1 |
NR_120577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009400.hg.1 |
NR_120577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009400.hg.1 |
NR_120577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009400.hg.1 |
NR_120577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009400.hg.1 |
NR_120579.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009400.hg.1 |
NR_120579.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009400.hg.1 |
NR_120579.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009400.hg.1 |
NR_120579.1 |
Refseq |