Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012157.hg.1 |
ENST00000536529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012157.hg.1 |
ENST00000536529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012157.hg.1 |
ENST00000536529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012157.hg.1 |
ENST00000536529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012157.hg.1 |
ENST00000536529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012157.hg.1 |
ENST00000536529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012157.hg.1 |
ENST00000536529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012157.hg.1 |
ENST00000536529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012157.hg.1 |
ENST00000536529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012157.hg.1 |
ENST00000536529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012157.hg.1 |
ENST00000538641.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012157.hg.1 |
ENST00000538641.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012157.hg.1 |
ENST00000538641.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012157.hg.1 |
ENST00000538641.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012157.hg.1 |
ENST00000545630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012157.hg.1 |
ENST00000545630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012157.hg.1 |
ENST00000545630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012157.hg.1 |
ENST00000545630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012157.hg.1 |
NR_120585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012157.hg.1 |
NR_120585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012157.hg.1 |
NR_120585.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012157.hg.1 |
NR_120585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012157.hg.1 |
ENST00000536529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012157.hg.1 |
ENST00000536529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012157.hg.1 |
ENST00000536529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012157.hg.1 |
ENST00000536529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012157.hg.1 |
ENST00000536529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012157.hg.1 |
ENST00000536529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012157.hg.1 |
ENST00000536529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012157.hg.1 |
ENST00000536529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012157.hg.1 |
ENST00000536529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012157.hg.1 |
ENST00000536529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012157.hg.1 |
ENST00000538641.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012157.hg.1 |
ENST00000538641.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012157.hg.1 |
ENST00000538641.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012157.hg.1 |
ENST00000538641.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012157.hg.1 |
ENST00000545630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012157.hg.1 |
ENST00000545630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012157.hg.1 |
ENST00000545630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012157.hg.1 |
ENST00000545630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012157.hg.1 |
NR_120585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012157.hg.1 |
NR_120585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012157.hg.1 |
NR_120585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012157.hg.1 |
NR_120585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193443.hg.1 |
ENST00000545630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193443.hg.1 |
NR_120585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193443.hg.1 |
ENST00000545630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193443.hg.1 |
NR_120585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193443.hg.1 |
TC1100012157.hg.1 |
Clariom D TC |