Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008127.hg.1 |
ENST00000428936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008127.hg.1 |
ENST00000428936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008127.hg.1 |
ENST00000428936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008127.hg.1 |
ENST00000428936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008127.hg.1 |
ENST00000430464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008127.hg.1 |
ENST00000430464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008127.hg.1 |
NR_120654.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008127.hg.1 |
NR_120654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000155148.hg.1 |
ENST00000428936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000155148.hg.1 |
ENST00000428936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000155148.hg.1 |
NR_120654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000155148.hg.1 |
ENST00000428936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000155148.hg.1 |
ENST00000428936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000155148.hg.1 |
NR_120654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000155148.hg.1 |
TC1000008127.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000008127.hg.1 |
ENST00000428936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008127.hg.1 |
ENST00000428936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008127.hg.1 |
ENST00000428936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008127.hg.1 |
ENST00000428936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008127.hg.1 |
ENST00000430464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008127.hg.1 |
ENST00000430464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008127.hg.1 |
NR_120654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008127.hg.1 |
NR_120654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000155151.hg.1 |
ENST00000428936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000155151.hg.1 |
ENST00000428936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000155151.hg.1 |
ENST00000430464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000155151.hg.1 |
NR_120654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000155151.hg.1 |
ENST00000428936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000155151.hg.1 |
ENST00000428936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000155151.hg.1 |
ENST00000430464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000155151.hg.1 |
NR_120654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000155151.hg.1 |
TC1000008127.hg.1 |
Clariom D TC |