Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011278.hg.1 |
ENST00000454178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011278.hg.1 |
ENST00000454178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011278.hg.1 |
ENST00000454178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011278.hg.1 |
ENST00000454178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011278.hg.1 |
ENST00000454178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011278.hg.1 |
ENST00000454178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011278.hg.1 |
NR_120667.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011278.hg.1 |
NR_120667.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011278.hg.1 |
NR_120667.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011278.hg.1 |
NR_120667.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011278.hg.1 |
NR_120667.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011278.hg.1 |
NR_120667.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011278.hg.1 |
NR_120668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011278.hg.1 |
NR_120668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011278.hg.1 |
NR_120668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011278.hg.1 |
NR_120668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011278.hg.1 |
NR_120668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011278.hg.1 |
NR_120668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000176742.hg.1 |
ENST00000454178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000176742.hg.1 |
NR_120667.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000176742.hg.1 |
NR_120668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000176742.hg.1 |
ENST00000454178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000176742.hg.1 |
NR_120667.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000176742.hg.1 |
NR_120668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000176742.hg.1 |
TC1000011278.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000176745.hg.1 |
NR_120668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000176745.hg.1 |
NR_120668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000176745.hg.1 |
TC1000011278.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000176744.hg.1 |
NR_120668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000176744.hg.1 |
NR_120668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000176744.hg.1 |
TC1000011278.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000011278.hg.1 |
ENST00000454178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011278.hg.1 |
ENST00000454178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011278.hg.1 |
ENST00000454178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011278.hg.1 |
ENST00000454178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011278.hg.1 |
ENST00000454178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011278.hg.1 |
ENST00000454178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011278.hg.1 |
NR_120667.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011278.hg.1 |
NR_120667.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011278.hg.1 |
NR_120667.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011278.hg.1 |
NR_120667.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011278.hg.1 |
NR_120667.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011278.hg.1 |
NR_120667.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011278.hg.1 |
NR_120668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011278.hg.1 |
NR_120668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011278.hg.1 |
NR_120668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011278.hg.1 |
NR_120668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011278.hg.1 |
NR_120668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011278.hg.1 |
NR_120668.1 |
Refseq |