Assay Details
Gene Symbol: LINC01474 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_121120.1 | 3 - 4 | 339 | 99 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transmembrane channel like 1 | TMC1 |
DFNA36, DFNB11, DFNB7 |
-35.4kb | Anti-sense | Chr. 9 - 72521801 - 72836351 on NCBI Build GRCh38 |
Downstream | aldehyde dehydrogenase 1 family member A1 | ALDH1A1 |
ALDC, ALDH-E1, ALDH1, ALDH11, HEL-9, HEL-S-53e, HEL12, PUMB1, RALDH1 |
28.4kb | Sense | Chr. 9 - 72900662 - 72953317 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000453787.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000453787.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000453787.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000453787.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000453787.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | ENST00000453787.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | NR_121120.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | NR_121120.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | NR_121120.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | NR_121120.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | NR_121120.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | NR_121120.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | NR_121120.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | NR_121180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | NR_121180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | NR_121180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | NR_121180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | NR_121180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | NR_121180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | NR_121180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | NR_121181.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | NR_121181.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | NR_121181.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | NR_121181.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | NR_121181.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012244.hg.1 | NR_121181.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900186200.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186200.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186200.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186200.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186200.hg.1 | ENST00000453787.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186200.hg.1 | NR_121120.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900186200.hg.1 | NR_121180.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900186200.hg.1 | NR_121181.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900186200.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186200.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186200.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186200.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186200.hg.1 | ENST00000453787.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186200.hg.1 | NR_121120.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900186200.hg.1 | NR_121180.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900186200.hg.1 | NR_121181.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900186200.hg.1 | TC0900012244.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000453787.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000453787.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000453787.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000453787.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000453787.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | ENST00000453787.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | NR_121120.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | NR_121120.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | NR_121120.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | NR_121120.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | NR_121120.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | NR_121120.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | NR_121120.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | NR_121180.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | NR_121180.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | NR_121180.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | NR_121180.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | NR_121180.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | NR_121180.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | NR_121180.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | NR_121181.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | NR_121181.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | NR_121181.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | NR_121181.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | NR_121181.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012244.hg.1 | NR_121181.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900186199.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186199.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186199.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186199.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186199.hg.1 | ENST00000453787.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186199.hg.1 | NR_121120.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900186199.hg.1 | NR_121180.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900186199.hg.1 | NR_121181.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900186199.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186199.hg.1 | ENST00000423171.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186199.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186199.hg.1 | ENST00000449235.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186199.hg.1 | ENST00000453787.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900186199.hg.1 | NR_121120.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900186199.hg.1 | NR_121180.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900186199.hg.1 | NR_121181.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900186199.hg.1 | TC0900012244.hg.1 | Clariom D TC |
