Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011772.hg.1 |
ENST00000550088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011772.hg.1 |
ENST00000550088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011772.hg.1 |
ENST00000550088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011772.hg.1 |
NR_121211.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011772.hg.1 |
NR_121211.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011772.hg.1 |
NR_121211.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011772.hg.1 |
NR_121211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191293.hg.1 |
ENST00000550088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191293.hg.1 |
NR_121211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191293.hg.1 |
ENST00000550088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191293.hg.1 |
NR_121211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191293.hg.1 |
TC1200011772.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200011772.hg.1 |
ENST00000550088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011772.hg.1 |
ENST00000550088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011772.hg.1 |
ENST00000550088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011772.hg.1 |
NR_121211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011772.hg.1 |
NR_121211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011772.hg.1 |
NR_121211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011772.hg.1 |
NR_121211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191294.hg.1 |
ENST00000550088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191294.hg.1 |
NR_121211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191294.hg.1 |
ENST00000550088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191294.hg.1 |
NR_121211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191294.hg.1 |
TC1200011772.hg.1 |
Clariom D TC |