Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009216.hg.1 |
ENST00000429224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009216.hg.1 |
ENST00000429224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009216.hg.1 |
ENST00000429224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009216.hg.1 |
ENST00000429224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009216.hg.1 |
ENST00000429224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009216.hg.1 |
ENST00000429224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009216.hg.1 |
ENST00000429224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009216.hg.1 |
ENST00000450304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009216.hg.1 |
ENST00000450304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009216.hg.1 |
ENST00000450304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009216.hg.1 |
NR_121577.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009216.hg.1 |
NR_121577.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009216.hg.1 |
NR_121577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162819.hg.1 |
ENST00000429224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162819.hg.1 |
NR_121577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162819.hg.1 |
ENST00000429224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162819.hg.1 |
NR_121577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162819.hg.1 |
TC0900009216.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900162836.hg.1 |
ENST00000450304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162836.hg.1 |
NR_121577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162836.hg.1 |
ENST00000450304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162836.hg.1 |
NR_121577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162836.hg.1 |
TC0900009216.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900162831.hg.1 |
ENST00000429224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162831.hg.1 |
ENST00000450304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162831.hg.1 |
NR_121577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162831.hg.1 |
ENST00000429224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162831.hg.1 |
ENST00000450304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162831.hg.1 |
NR_121577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162831.hg.1 |
TC0900009216.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900009216.hg.1 |
ENST00000429224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009216.hg.1 |
ENST00000429224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009216.hg.1 |
ENST00000429224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009216.hg.1 |
ENST00000429224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009216.hg.1 |
ENST00000429224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009216.hg.1 |
ENST00000429224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009216.hg.1 |
ENST00000429224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009216.hg.1 |
ENST00000450304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009216.hg.1 |
ENST00000450304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009216.hg.1 |
ENST00000450304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009216.hg.1 |
NR_121577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009216.hg.1 |
NR_121577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009216.hg.1 |
NR_121577.1 |
Refseq |