Assay Details
Gene Symbol: LOC101928786 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_121582.1 | 1 - 2 | 192 | 118 | ||
GenBank mRNA | AK126935.1 | - | 1 - 2 | 192 | 118 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC105376331 | LOC105376331 | -96.4kb | Sense | Chr. 9 - 137943630 - 137947060 on NCBI Build GRCh38 | |
Downstream | tubulin beta pseudogene 5 | TUBBP5 | 105.0kb | Anti-sense | Chr. 9 - 138150113 - 138177433 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
