Assay Details
| Gene Symbol: BGLT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_121648.1 | 1 | 120 | 91 | ||
| GenBank mRNA | AY034471.1 | - | 1 | 120 | 91 | |
| BU658638.1 | - | 1 | 130 | 91 | ||
| KF110790.1 | - | 1 | 2797 | 91 | ||
More Information
Set Membership: |
|

BGLT3
Genomic Map