Assay Details
| Gene Symbol: LOC101927296 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_121660.1 | 1 - 2 | 322 | 172 | ||
| GenBank mRNA | BI767720.1 | - | 1 - 2 | 322 | 172 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | protein phosphatase 4 regulatory subunit 2 | PPP4R2 | PP4R2 | -455.5kb | Sense | Chr. 3 - 72996743 - 73069201 on NCBI Build GRCh38 |
| Downstream | PDZRN3 antisense RNA 1 | PDZRN3-AS1 | 53.8kb | Sense | Chr. 3 - 73623568 - 73627899 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101927296
Genomic Map