Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
ENST00000436032.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
ENST00000436032.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
ENST00000436032.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
ENST00000450920.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
ENST00000450920.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
ENST00000450920.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
ENST00000600323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
ENST00000600323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
ENST00000600323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
ENST00000600323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
ENST00000600323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
ENST00000600323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
ENST00000600323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
ENST00000600323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
ENST00000600323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
ENST00000600323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
ENST00000612302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
ENST00000612302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
ENST00000612302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
ENST00000612302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
ENST00000614304.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
ENST00000614304.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
ENST00000614304.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
ENST00000614304.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
ENST00000614304.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
ENST00000614304.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
NR_121663.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
NR_121663.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
NR_121663.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
NR_121663.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
NR_121663.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
NR_121664.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
NR_121664.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
NR_121664.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006538.hg.1 |
NR_121664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144855.hg.1 |
ENST00000450920.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144855.hg.1 |
NR_121663.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144855.hg.1 |
NR_121664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144855.hg.1 |
ENST00000450920.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144855.hg.1 |
NR_121663.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144855.hg.1 |
NR_121664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144855.hg.1 |
TC0300006538.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300144853.hg.1 |
ENST00000450920.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144853.hg.1 |
ENST00000600323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144853.hg.1 |
ENST00000600323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144853.hg.1 |
NR_121663.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144853.hg.1 |
NR_121664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144853.hg.1 |
ENST00000450920.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144853.hg.1 |
ENST00000600323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144853.hg.1 |
ENST00000600323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144853.hg.1 |
NR_121663.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144853.hg.1 |
NR_121664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144853.hg.1 |
TC0300006538.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
ENST00000436032.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
ENST00000436032.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
ENST00000436032.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
ENST00000450920.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
ENST00000450920.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
ENST00000450920.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
ENST00000600323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
ENST00000600323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
ENST00000600323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
ENST00000600323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
ENST00000600323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
ENST00000600323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
ENST00000600323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
ENST00000600323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
ENST00000600323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
ENST00000600323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
ENST00000612302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
ENST00000612302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
ENST00000612302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
ENST00000612302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
ENST00000614304.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
ENST00000614304.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
ENST00000614304.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
ENST00000614304.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
ENST00000614304.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
ENST00000614304.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
NR_121663.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
NR_121663.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
NR_121663.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
NR_121663.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
NR_121663.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
NR_121664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
NR_121664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
NR_121664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006538.hg.1 |
NR_121664.1 |
Refseq |