Assay Details
Gene Symbol: ERICH3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_121670.1 | 2 - 3 | 267 | 86 | ||
NR_121671.1 | 2 - 3 | 174 | 86 | |||
GenBank mRNA | BX092187.1 | - | 2 - 3 | 267 | 86 | |
CR627203.1 | - | 2 - 3 | 174 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | FPGT-TNNI3K readthrough | FPGT-TNNI3K | -70.8kb | Sense | Chr. 1 - 74198212 - 74544432 on NCBI Build GRCh38 | |
Downstream | crystallin zeta | CRYZ | 79.4kb | Anti-sense | Chr. 1 - 74705482 - 74733408 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000433746.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000433746.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000433746.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000614534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000614534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000614534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | ENST00000416017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | ENST00000416017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | ENST00000416017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | ENST00000416017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | ENST00000416017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | ENST00000416017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | ENST00000612390.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | ENST00000612390.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | ENST00000612390.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | ENST00000612390.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | ENST00000612390.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | ENST00000612390.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | ENST00000612390.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | ENST00000620678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | ENST00000620678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | NR_121671.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | NR_121671.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | NR_121671.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | NR_121671.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | NR_121671.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | NR_121671.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008750.hg.1 | NR_121671.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | ENST00000416017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | ENST00000416017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | ENST00000416017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | ENST00000416017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | ENST00000416017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | ENST00000416017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | ENST00000612390.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | ENST00000612390.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | ENST00000612390.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | ENST00000612390.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | ENST00000612390.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | ENST00000612390.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | ENST00000612390.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | ENST00000620678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | ENST00000620678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | NR_121671.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | NR_121671.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | NR_121671.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | NR_121671.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | NR_121671.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | NR_121671.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008750.hg.1 | NR_121671.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100162324.hg.1 | ENST00000416017.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100162324.hg.1 | ENST00000620678.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100162324.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100162324.hg.1 | ENST00000416017.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100162324.hg.1 | ENST00000620678.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100162324.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100162324.hg.1 | TC0100008750.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100162331.hg.1 | ENST00000612390.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100162331.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100162331.hg.1 | NR_121671.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100162331.hg.1 | ENST00000612390.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100162331.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100162331.hg.1 | NR_121671.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100162331.hg.1 | TC0100008750.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100162329.hg.1 | ENST00000416017.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100162329.hg.1 | ENST00000612390.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100162329.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100162329.hg.1 | NR_121671.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100162329.hg.1 | ENST00000416017.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100162329.hg.1 | ENST00000612390.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100162329.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100162329.hg.1 | NR_121671.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100162329.hg.1 | TC0100008750.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100162327.hg.1 | ENST00000416017.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100162327.hg.1 | ENST00000612390.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100162327.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100162327.hg.1 | NR_121671.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100162327.hg.1 | ENST00000416017.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100162327.hg.1 | ENST00000612390.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100162327.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100162327.hg.1 | NR_121671.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100162327.hg.1 | TC0100008750.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100162328.hg.1 | ENST00000416017.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100162328.hg.1 | ENST00000612390.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100162328.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100162328.hg.1 | NR_121671.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100162328.hg.1 | ENST00000416017.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100162328.hg.1 | ENST00000612390.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100162328.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100162328.hg.1 | NR_121671.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100162328.hg.1 | TC0100008750.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100162330.hg.1 | ENST00000612390.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100162330.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100162330.hg.1 | NR_121671.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100162330.hg.1 | ENST00000612390.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100162330.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100162330.hg.1 | NR_121671.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100162330.hg.1 | TC0100008750.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100162323.hg.1 | ENST00000416017.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100162323.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100162323.hg.1 | ENST00000416017.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100162323.hg.1 | NR_121670.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100162323.hg.1 | TC0100008750.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000433746.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000433746.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000433746.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000614534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000614534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000614534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
