Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000507318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000507318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000507318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000507318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000507318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000507318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000507373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000507373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000509139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000509139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000509139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000509139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000509139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000512007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000512007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000515304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000515304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000515304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
NR_122044.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
NR_122044.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
NR_122044.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
NR_122044.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
NR_122044.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009053.hg.1 |
NR_122044.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500163066.hg.1 |
ENST00000509139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163066.hg.1 |
NR_122044.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500163066.hg.1 |
ENST00000509139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163066.hg.1 |
NR_122044.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500163066.hg.1 |
TC0500009053.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000507318.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000507318.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000507318.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000507318.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000507318.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000507318.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000507373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000507373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000509139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000509139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000509139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000509139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000509139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000512007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000512007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000515304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000515304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000515304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
ENST00000515519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
NR_122044.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
NR_122044.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
NR_122044.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
NR_122044.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
NR_122044.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009053.hg.1 |
NR_122044.1 |
Refseq |