Assay Details
| Gene Symbol: ACTA2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125373.1 | 2 - 3 | 525 | 80 | ||
| GenBank mRNA | AX748062.1 | - | 2 - 3 | 525 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | STAM binding protein like 1 | STAMBPL1 |
ALMalpha, AMSH-FP, AMSH-LP, bA399O19.2 |
-10.1kb | Sense | Chr. 10 - 88879368 - 88923502 on NCBI Build GRCh38 |
| Downstream | Fas cell surface death receptor | FAS |
ALPS1A, APO-1, APT1, CD95, FAS1, FASTM, TNFRSF6 |
34.2kb | Sense | Chr. 10 - 88968429 - 89017061 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000596007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000596007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000596007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000596007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000596007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000625277.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000625277.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000625277.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000625277.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000627669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000627669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000627669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000627669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | NR_125373.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | NR_125373.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | NR_125373.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | NR_125373.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | NR_125373.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | NR_125373.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | NR_125373.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | NR_125373.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | NR_125373.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000224784.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000224784.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000224784.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000224784.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000224784.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000224784.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000224784.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000224784.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000224784.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000415557.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000415557.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000415557.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000415557.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000415557.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000415557.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000415557.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000458159.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000458159.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000458159.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000458159.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000458159.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000458208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000458208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000458208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000458208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000458208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000458208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000458208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000458208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000458208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000458208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000458208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000458208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000458208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000480297.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000480297.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000480297.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000480297.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000480297.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000480297.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000482085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000482085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000488967.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000488967.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | ENST00000488967.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | NM_001141945.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | NM_001141945.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | NM_001141945.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | NM_001141945.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | NM_001141945.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | NM_001141945.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | NM_001141945.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | NM_001141945.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | NM_001141945.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | NM_001141945.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | NM_001141945.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | NM_001141945.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | NM_001141945.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | NM_001613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | NM_001613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | NM_001613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | NM_001613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | NM_001613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | NM_001613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | NM_001613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | NM_001613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | NM_001613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011370.hg.1 | NM_001613.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156839.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156839.hg.1 | NR_125373.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156839.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156839.hg.1 | NR_125373.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156839.hg.1 | TC1000008387.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000156837.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156837.hg.1 | NR_125373.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156837.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156837.hg.1 | NR_125373.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156837.hg.1 | TC1000008387.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000596007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000596007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000596007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000596007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000596007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000625277.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000625277.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000625277.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000625277.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000627669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000627669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000627669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000627669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | NR_125373.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | NR_125373.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | NR_125373.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | NR_125373.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | NR_125373.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | NR_125373.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | NR_125373.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | NR_125373.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | NR_125373.1 | Refseq |

ACTA2-AS1
Genomic Map