Assay Details
Gene Symbol: ACTA2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125373.1 | 4 - 5 | 1462 | 85 | ||
GenBank mRNA | AX748062.1 | - | 4 - 5 | 1462 | 85 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | STAM binding protein like 1 | STAMBPL1 |
ALMalpha, AMSH-FP, AMSH-LP, bA399O19.2 |
-14.7kb | Sense | Chr. 10 - 88879368 - 88923502 on NCBI Build GRCh38 |
Downstream | Fas cell surface death receptor | FAS |
ALPS1A, APO-1, APT1, CD95, FAS1, FASTM, TNFRSF6 |
29.8kb | Sense | Chr. 10 - 88968429 - 89017061 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000596007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000596007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000596007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000596007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000596007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000625277.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000625277.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000625277.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000625277.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000627669.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000627669.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000627669.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | ENST00000627669.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | NR_125373.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | NR_125373.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | NR_125373.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | NR_125373.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | NR_125373.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | NR_125373.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | NR_125373.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | NR_125373.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008387.hg.1 | NR_125373.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156843.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156843.hg.1 | ENST00000596007.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156843.hg.1 | ENST00000625277.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156843.hg.1 | NR_125373.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156843.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156843.hg.1 | ENST00000596007.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156843.hg.1 | ENST00000625277.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156843.hg.1 | NR_125373.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156843.hg.1 | TC1000008387.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000156845.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156845.hg.1 | NR_125373.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156845.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156845.hg.1 | NR_125373.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156845.hg.1 | TC1000008387.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000156844.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156844.hg.1 | ENST00000596007.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156844.hg.1 | NR_125373.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156844.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156844.hg.1 | ENST00000596007.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156844.hg.1 | NR_125373.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156844.hg.1 | TC1000008387.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000156841.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156841.hg.1 | NR_125373.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156841.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156841.hg.1 | NR_125373.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156841.hg.1 | TC1000008387.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000437930.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000596007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000596007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000596007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000596007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000596007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000625277.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000625277.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000625277.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000625277.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000627669.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000627669.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000627669.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | ENST00000627669.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | NR_125373.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | NR_125373.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | NR_125373.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | NR_125373.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | NR_125373.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | NR_125373.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | NR_125373.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | NR_125373.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008387.hg.1 | NR_125373.1 | Refseq |
